Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO456793153

Apollo Scientific Ltd

1-Bromo-4-(tert-butoxy)benzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
229.12
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

1-bromo-4-tert-butoxybenzene

Inchi Code

1S/C10H13BrO/c1-10(2,3)12-9-6-4-8(11)5-7-9/h4-7H,1-3H3

InChI key

QIWQHUCUWNGYDZ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/1-bromo-4-tert-butoxybenzene

Flash Point

63-65/0.4mm

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/1-bromo-4-tert-butoxybenzene


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service