APO455838442
Apollo Scientific Ltd
2-Fluoro-4-(trifluoromethyl)phenacyl bromide
About This Item
bp
30-37
Iupac Name
2-bromo-1-[2-fluoro-4-(trifluoromethyl)phenyl]ethanone
Inchi Code
1S/C9H5BrF4O/c10-4-8(15)6-2-1-5(3-7(6)11)9(12,13)14/h1-3H,4H2
InChI key
NHVLVTWQEUHHBM-UHFFFAOYSA-N
storage temp.
Country of Origin
Shipping Temperature
Physical Form
Product Link
Synonym
Boiling point
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/2-fluoro-4-trifluoromethylphenacyl-bromide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service