Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455838234

Apollo Scientific Ltd

5,5,5-Trifluoro-DL-norvaline

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$356.40
250 MG
$618.20

Vendor SKU: PC7813-100MG

$356.40


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$356.40
250 MG
$618.20

About This Item

CAS Number:
Molecular Weight:
171.12
MDL number:

Vendor SKU: PC7813-100MG

$356.40


Ships in 7 days - from Aldrich Partner.

bp

256-258

mp

>110

Iupac Name

5,5,5-trifluoronorvaline

Inchi Code

1S/C5H8F3NO2/c6-5(7,8)2-1-3(9)4(10)11/h3H,1-2,9H2,(H,10,11)

InChI key

KTZZIDWVKLDWBF-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

96%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/555-trifluoro-dl-norvaline

Synonym

2-Amino-5,5,5-trifluoropentanoic acid, 2-Amino-5,5,5-trifluorovaleric acid

Boiling point

256-258

Melting Point

>110

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/555-trifluoro-dl-norvaline


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service