APO455838019
Apollo Scientific Ltd
2-(Trifluoromethoxy)benzonitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
density
1.245
Iupac Name
2-(trifluoromethoxy)benzonitrile
Inchi Code
1S/C8H4F3NO/c9-8(10,11)13-7-4-2-1-3-6(7)5-12/h1-4H
InChI key
ACNBBQGAWMHXLA-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
97%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Liquid
Product Link
https://store.apolloscientific.co.uk/product/2-trifluoromethoxybenzonitrile
Flash Point
61-62/8mm
density
1.245
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/2-trifluoromethoxybenzonitrile
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service