Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455837917

Apollo Scientific Ltd

3,4,5-Trifluorobenzophenone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
236.19
MDL number:

bp

77-78

Iupac Name

phenyl(3,4,5-trifluorophenyl)methanone

Inchi Code

1S/C13H7F3O/c14-10-6-9(7-11(15)12(10)16)13(17)8-4-2-1-3-5-8/h1-7H

InChI key

BTQOMZVQASIULU-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Product Link

https://store.apolloscientific.co.uk/product/345-trifluorobenzophenone

Boiling point

77-78

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/345-trifluorobenzophenone


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service