APO455837449
Apollo Scientific Ltd
4-Bromo-3-(trifluoromethyl)benzyl bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
52-53
Iupac Name
1-bromo-4-(bromomethyl)-2-(trifluoromethyl)benzene
Inchi Code
1S/C8H5Br2F3/c9-4-5-1-2-7(10)6(3-5)8(11,12)13/h1-3H,4H2
Inchi Key
UHHXITPNGSCOBA-UHFFFAOYSA-N
Storage temp.
Ambient Temperature
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/4-bromo-3-trifluoromethylbenzyl-bromide
Synonym
1-Bromo-4-(bromomethyl)-2-(trifluoromethyl)benzene, 2-Bromo-5-(bromomethyl)benzotrifluoride
Boiling point
52-53
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/4-bromo-3-trifluoromethylbenzyl-bromide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service