APO455837260
Apollo Scientific Ltd
N-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
About This Item
bp
65-66
Iupac Name
1-[2-nitro-4-(trifluoromethyl)phenyl]piperazine
Inchi Code
1S/C11H12F3N3O2/c12-11(13,14)8-1-2-9(10(7-8)17(18)19)16-5-3-15-4-6-16/h1-2,7,15H,3-6H2
InChI key
YOBUPGXTLFRIJD-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Synonym
Boiling point
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/n-2-nitro-4-trifluoromethylphenylpiperazine
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service