Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455837065

Apollo Scientific Ltd

4-(Trifluoromethoxy)benzene-1,2-diamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
192.14
MDL number:

mp

>110

Iupac Name

4-(trifluoromethoxy)-1,2-benzenediamine

Inchi Code

1S/C7H7F3N2O/c8-7(9,10)13-4-1-2-5(11)6(12)3-4/h1-3H,11-12H2

InChI key

PIOKGAUSPFWRMD-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/4-trifluoromethoxybenzene-12-diamine

Synonym

1,2-Diamino-4-(trifluoromethoxy)benzene

Flash Point

106-108/5mm

Melting Point

>110

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-trifluoromethoxybenzene-12-diamine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service