Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455836305

Apollo Scientific Ltd

3'-Fluoro-5'-(trifluoromethyl)propiophenone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$22.00
5 G
$89.10
25 G
$276.10

Vendor SKU: PC4377T-1G

$22.00


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$22.00
5 G
$89.10
25 G
$276.10

About This Item

CAS Number:
Molecular Weight:
220.17
MDL number:

Vendor SKU: PC4377T-1G

$22.00


Ships in 7 days - from Aldrich Partner.

Iupac Name

1-[3-fluoro-5-(trifluoromethyl)phenyl]-1-propanone

Inchi Code

1S/C10H8F4O/c1-2-9(15)6-3-7(10(12,13)14)5-8(11)4-6/h3-5H,2H2,1H3

Inchi Key

KEAIAOPGXGEINJ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/3-fluoro-5-trifluoromethylpropiophenone

Synonym

1-[3-Fluoro-5-(trifluoromethyl)phenyl]propan-1-one, 3-Fluoro-5-propanoylbenzotrifluoride

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-fluoro-5-trifluoromethylpropiophenone


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service