Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455835737

Apollo Scientific Ltd

Ethyl 4,4,4-trifluoro-3-(trifluoromethyl)crotonate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$160.60

Vendor SKU: PC3289G-5G

$160.60


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
5 G
$160.60

About This Item

CAS Number:
Molecular Weight:
236.11
MDL number:

Vendor SKU: PC3289G-5G

$160.60


Ships in 7 days - from Aldrich Partner.

mp

46

density

1.3525

Iupac Name

ethyl 4,4,4-trifluoro-3-(trifluoromethyl)-2-butenoate

Inchi Code

1S/C7H6F6O2/c1-2-15-5(14)3-4(6(8,9)10)7(11,12)13/h3H,2H2,1H3

InChI key

CULZFNOUPBWSIZ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/ethyl-444-trifluoro-3-trifluoromethylcrotonate

Synonym

Ethyl 4,4,4-trifluoro-3-(trifluoromethyl)but-2-enoate

Flash Point

127-128

Melting Point

46

density

1.3525

pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/ethyl-444-trifluoro-3-trifluoromethylcrotonate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service