Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455835430

Apollo Scientific Ltd

Methyl 2-fluoro-2-methylpropanoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
120.12
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

22

density

1

Iupac Name

methyl 2-fluoro-2-methylpropanoate

Inchi Code

1S/C5H9FO2/c1-5(2,6)4(7)8-3/h1-3H3

Inchi Key

OEGBOFOVYSOERL-UHFFFAOYSA-N

Storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/methyl-2-fluoro-2-methylpropanoate

Synonym

Methyl 2-fluoro-2-methylpropionate

Flash Point

108-109

Melting Point

22

density

1

Pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/methyl-2-fluoro-2-methylpropanoate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service