Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455835424

Apollo Scientific Ltd

5-Bromo-2-fluoroanisole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
205.03
MDL number:

mp

98

Iupac Name

5-bromo-2-fluorophenyl methyl ether

Inchi Code

1S/C7H6BrFO/c1-10-7-4-5(8)2-3-6(7)9/h2-4H,1H3

InChI key

SEVMQEIGENUPIE-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/5-bromo-2-fluoroanisole

Synonym

4-Bromo-1-fluoro-2-methoxybenzene, 5-Bromo-2-fluorophenyl methyl ether

Flash Point

82-85

Melting Point

98

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/5-bromo-2-fluoroanisole


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service