APO455835361
Apollo Scientific Ltd
1-(Difluoromethoxy)-4-iodobenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
110
density
1.871
Iupac Name
difluoromethyl 4-iodophenyl ether
Inchi Code
1S/C7H5F2IO/c8-7(9)11-6-3-1-5(10)2-4-6/h1-4,7H
InChI key
KBNPQVHJUYEJIZ-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
98%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Liquid
Product Link
https://store.apolloscientific.co.uk/product/1-difluoromethoxy-4-iodobenzene
Synonym
Difluoromethyl 4-iodophenyl ether, alpha,alpha-Difluoro-4-iodoanisole
Flash Point
217-219
Melting Point
110
density
1.871
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/1-difluoromethoxy-4-iodobenzene
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service