APO455835342
Apollo Scientific Ltd
4-Ethyl-5-fluoro-6-hydroxypyrimidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
6-ethyl-5-fluoro-4(3H)-pyrimidinone
Inchi Code
1S/C6H7FN2O/c1-2-4-5(7)6(10)9-3-8-4/h3H,2H2,1H3,(H,8,9,10)
InChI key
ZEMRCKIJEFNNCO-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/4-ethyl-5-fluoro-6-hydroxypyrimidine
Synonym
6-Ethyl-5-fluoropyrimidin-4-ol
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/4-ethyl-5-fluoro-6-hydroxypyrimidine
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service