Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455834642

Apollo Scientific Ltd

2-Fluoro-5-methylbenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
138.14
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

81

density

1.129

Iupac Name

2-fluoro-5-methylbenzaldehyde

Inchi Code

1S/C8H7FO/c1-6-2-3-8(9)7(4-6)5-10/h2-5H,1H3

Inchi Key

QAWILFXCNRBMDF-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/2-fluoro-5-methylbenzaldehyde

Synonym

4-Fluoro-3-formyltoluene, 6-Fluoro-m-tolualdehyde

Flash Point

89/15mm

Melting Point

81

density

1.129

pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-fluoro-5-methylbenzaldehyde


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service