Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455834184

Apollo Scientific Ltd

Butyl perfluorobutanoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
270.15
MDL number:

density

1.296

Iupac Name

butyl 2,2,3,3,4,4,4-heptafluorobutanoate

Inchi Code

1S/C8H9F7O2/c1-2-3-4-17-5(16)6(9,10)7(11,12)8(13,14)15/h2-4H2,1H3

InChI key

YDXXJZKOOOJVEE-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/butyl-perfluorobutanoate

Synonym

Butyl heptafluorobutanoate, Butyl heptafluorobutyrate

Flash Point

131-133/740mm

density

1.296

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/butyl-perfluorobutanoate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service