Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455832864

Apollo Scientific Ltd

1-Acetyl-5-aminoindoline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
176.22
MDL number:

bp

181-184

Iupac Name

1-acetyl-2,3-dihydro-1H-indol-5-ylamine

Inchi Code

1S/C10H12N2O/c1-7(13)12-5-4-8-6-9(11)2-3-10(8)12/h2-3,6H,4-5,11H2,1H3

InChI key

WSDUFDGEYKOQRT-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/1-acetyl-5-aminoindoline

Boiling point

181-184

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/1-acetyl-5-aminoindoline


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service