APO455832029
Apollo Scientific Ltd
2,5-Dichlorobenzoyl chloride
About This Item
bp
23
Iupac Name
2,5-dichlorobenzoyl chloride
Inchi Code
1S/C7H3Cl3O/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3H
InChI key
RSINFFVFOTUDEC-UHFFFAOYSA-N
storage temp.
Country of Origin
Shipping Temperature
Physical Form
Product Link
Flash Point
Boiling point
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/25-dichlorobenzoyl-chloride
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service