Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455831641

Apollo Scientific Ltd

3-Amino-5-(tert-butyl)thiophene-2-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$28.60

Vendor SKU: OR5325-100MG

$28.60


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$28.60

About This Item

CAS Number:
Molecular Weight:
198.29
MDL number:

Vendor SKU: OR5325-100MG

$28.60


Ships in 7 days - from Aldrich Partner.

bp

146-148

Iupac Name

3-amino-5-tert-butyl-2-thiophenecarboxamide

Inchi Code

1S/C9H14N2OS/c1-9(2,3)6-4-5(10)7(13-6)8(11)12/h4H,10H2,1-3H3,(H2,11,12)

InChI key

MKHZJBZWKNDDCN-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/3-amino-5-tert-butylthiophene-2-carboxamide

Boiling point

146-148

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-amino-5-tert-butylthiophene-2-carboxamide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service