APO455830683
Apollo Scientific Ltd
1,3-Dimethyl-1H-indazole-6-boronic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1,3-dimethyl-1H-indazol-6-ylboronic acid
Inchi Code
1S/C9H11BN2O2/c1-6-8-4-3-7(10(13)14)5-9(8)12(2)11-6/h3-5,13-14H,1-2H3
InChI key
FOUSXGGKIFZVIQ-UHFFFAOYSA-N
storage temp.
Store at 2-8C
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid-Powder
Product Link
https://store.apolloscientific.co.uk/product/13-dimethyl-1h-indazole-6-boronic-acid
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/13-dimethyl-1h-indazole-6-boronic-acid
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service