APO455829350
Apollo Scientific Ltd
2-(Morpholin-4-yl)acetamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
122
Iupac Name
2-(4-morpholinyl)acetamide
Inchi Code
1S/C6H12N2O2/c7-6(9)5-8-1-3-10-4-2-8/h1-5H2,(H2,7,9)
InChI key
BWRNWWSQZROEOA-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/2-morpholin-4-ylacetamide
Boiling point
122
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/2-morpholin-4-ylacetamide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service