APO455828439
Apollo Scientific Ltd
5-Methylfuran-2-boronic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
82
Iupac Name
5-methyl-2-furylboronic acid
Inchi Code
1S/C5H7BO3/c1-4-2-3-5(9-4)6(7)8/h2-3,7-8H,1H3
InChI key
LMJIDBVSSSPRII-UHFFFAOYSA-N
storage temp.
Store at 2-8C
Purity
97%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/5-methylfuran-2-boronic-acid
Boiling point
82
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/5-methylfuran-2-boronic-acid
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service