Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455827009

Apollo Scientific Ltd

3-[(Pyrrolidin-3-yl)methyl]pyridine oxalate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$37.40

Vendor SKU: OR12929-50MG

$37.40


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
50 MG
$37.40

About This Item

CAS Number:
Molecular Weight:
252.27
MDL number:

Vendor SKU: OR12929-50MG

$37.40


Ships in 7 days - from Aldrich Partner.

Iupac Name

3-(3-pyrrolidinylmethyl)pyridine oxalate

Inchi Code

1S/C10H14N2.C2H2O4/c1-2-9(7-11-4-1)6-10-3-5-12-8-10;3-1(4)2(5)6/h1-2,4,7,10,12H,3,5-6,8H2;(H,3,4)(H,5,6)

InChI key

IMVHZHYTGHDXCA-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/3-pyrrolidin-3-ylmethylpyridine-oxalate

pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-pyrrolidin-3-ylmethylpyridine-oxalate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service