Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO455825951

Apollo Scientific Ltd

4,6-Dimethylpyrimidine-2-carbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$127.60

Vendor SKU: OR0829-1G

$127.60


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$127.60

About This Item

CAS Number:
Molecular Weight:
133.15
MDL number:

Vendor SKU: OR0829-1G

$127.60


Ships in 7 days - from Aldrich Partner.

bp

82-83

Iupac Name

4,6-dimethyl-2-pyrimidinecarbonitrile

Inchi Code

1S/C7H7N3/c1-5-3-6(2)10-7(4-8)9-5/h3H,1-2H3

InChI key

UJUJKEOTNKJDHF-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/46-dimethylpyrimidine-2-carbonitrile

Boiling point

82-83

pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/46-dimethylpyrimidine-2-carbonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service