Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO428499078

Apollo Scientific Ltd

3-Fluoro-2-(trifluoromethyl)phenylacetonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$176.00
5 G
$704.00
10 G
$1,195.70

Vendor SKU: PC302954-1G

$176.00


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 G
$176.00
5 G
$704.00
10 G
$1,195.70

About This Item

CAS Number:
Molecular Weight:
203.14
MDL number:

Vendor SKU: PC302954-1G

$176.00


Ships in 7 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

bp

27-29

Iupac Name

[3-fluoro-2-(trifluoromethyl)phenyl]acetonitrile

Inchi Code

1S/C9H5F4N/c10-7-3-1-2-6(4-5-14)8(7)9(11,12)13/h1-3H,4H2

InChI key

UADIGAUUNBZPDF-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/3-fluoro-2-trifluoromethylphenylacetonitrile

Boiling point

27-29

pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-fluoro-2-trifluoromethylphenylacetonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service