Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO428498297

Apollo Scientific Ltd

4-Methoxy-3-(trifluoromethyl)cinnamic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
246.19
MDL number:

bp

187-189

Iupac Name

(2E)-3-[4-methoxy-3-(trifluoromethyl)phenyl]-2-propenoic acid

Inchi Code

1S/C11H9F3O3/c1-17-9-4-2-7(3-5-10(15)16)6-8(9)11(12,13)14/h2-6H,1H3,(H,15,16)/b5-3+

InChI key

IOINNBJLNHLSHD-HWKANZROSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid-Viscous

Product Link

https://store.apolloscientific.co.uk/product/4-methoxy-3-trifluoromethylcinnamic-acid

Boiling point

187-189

pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-methoxy-3-trifluoromethylcinnamic-acid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service