Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO428497968

Apollo Scientific Ltd

2-[2-Fluoro-5-(trifluoromethyl)phenyl]ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$105.60
5 G
$441.10
25 G
$1,985.50
100 G
$5,955.40

Vendor SKU: PC302019-1G

$105.60


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
1 G
$105.60
5 G
$441.10
25 G
$1,985.50
100 G
$5,955.40

About This Item

CAS Number:
Molecular Weight:
208.16
MDL number:

Vendor SKU: PC302019-1G

$105.60


Ships in 7 days - from Aldrich Partner.

Iupac Name

2-[2-fluoro-5-(trifluoromethyl)phenyl]ethanol

Inchi Code

1S/C9H8F4O/c10-8-2-1-7(9(11,12)13)5-6(8)3-4-14/h1-2,5,14H,3-4H2

InChI key

YYIXBCMBMSZIOV-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/2-2-fluoro-5-trifluoromethylphenylethanol

pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-2-fluoro-5-trifluoromethylphenylethanol


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service