Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO414281866

Apollo Scientific Ltd

2-Methyl-1H-imidazole-4,5-dicarbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
132.12
MDL number:
Pricing and availability is not currently available. Contact Technical Service

bp

228-230

Iupac Name

2-methyl-1H-imidazole-4,5-dicarbonitrile

Inchi Code

1S/C6H4N4/c1-4-9-5(2-7)6(3-8)10-4/h1H3,(H,9,10)

InChI key

ZEAVNOQDMHVDLU-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/2-methyl-1h-imidazole-45-dicarbonitrile

Boiling point

228-230

pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-methyl-1h-imidazole-45-dicarbonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service