Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

APO414278813

Apollo Scientific Ltd

3-Bromo-5-chloro-2-hydroxybenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
237.48
MDL number:
Pricing and availability is not currently available. Contact Technical Service

bp

64-66

Iupac Name

2-bromo-4-chloro-6-(hydroxymethyl)phenol

Inchi Code

1S/C7H6BrClO2/c8-6-2-5(9)1-4(3-10)7(6)11/h1-2,10-11H,3H2

InChI key

RVRSNICKUNKOQP-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/3-bromo-5-chloro-2-hydroxybenzyl-alcohol

Boiling point

64-66

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-bromo-5-chloro-2-hydroxybenzyl-alcohol


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service