Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AOBH7BADDF37

AOBChem

1,4-Difluoro-2,3-dimethoxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$225.40
500 MG
$322.00
1 G
$450.80

Vendor SKU: 15869-250MG

$225.40


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$225.40
500 MG
$322.00
1 G
$450.80

About This Item

CAS Number:
Molecular Weight:
174.15
MDL number:

Vendor SKU: 15869-250MG

$225.40


Ships in 7 days - from Aldrich Partner.

Iupac Name

1,4-difluoro-2,3-dimethoxybenzene

Inchi Code

1S/C8H8F2O2/c1-11-7-5(9)3-4-6(10)8(7)12-2/h3-4H,1-2H3

InChI key

OUUAUECUXZLMQR-UHFFFAOYSA-N

Purity

97

Country of Origin

CN

Physical Form

Liquid

Product Link

https://aobchem.com/1-4-difluoro-2-3-dimethoxybenzene.html

MSDS

https://aobchem.com/pub/media/files/sds/15869.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service