Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH99C0345B

Ambeed, Inc.

4-Chloro-2-(difluoromethyl)pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$103.50
1 G
$211.60
5 G
$730.25

Vendor SKU: A423942-250MG

$103.50


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$103.50
1 G
$211.60
5 G
$730.25

About This Item

CAS Number:
Molecular Weight:
163.55
MDL number:

Vendor SKU: A423942-250MG

$103.50


Ships in 7 days - from Aldrich Partner.

Iupac Name

4-chloro-2-(difluoromethyl)pyridine

Inchi Code

1S/C6H4ClF2N/c7-4-1-2-10-5(3-4)6(8)9/h1-3,6H

InChI key

IYZZTRHKWXUPDQ-UHFFFAOYSA-N

storage temp.

Inert atmosphere,2-8C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Solid or Semi-solid or liquid

Product Link

https://www.ambeed.com/products/1193104-11-8.html

pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service