Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH996F256A

Ambeed, Inc.

Phenanthren-4-ol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$74.75
250 MG
$127.65
1 G
$331.20

Vendor SKU: A128613-100MG

$74.75


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$74.75
250 MG
$127.65
1 G
$331.20

About This Item

CAS Number:
Molecular Weight:
194.23
MDL number:

Vendor SKU: A128613-100MG

$74.75


Ships in 7 days - from Aldrich Partner.

Iupac Name

4-phenanthrenol

Inchi Code

1S/C14H10O/c15-13-7-3-5-11-9-8-10-4-1-2-6-12(10)14(11)13/h1-9,15H

InChI key

SIMYIUXARJLHEA-UHFFFAOYSA-N

storage temp.

Sealed in dry,Room Temperature

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/7651-86-7.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service