Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH98C8E5ED

Ambeed, Inc.

Ethyl 2-aminothiazole-4-carboxylate hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
253.12
MDL number:

Iupac Name

ethyl 2-amino-1,3-thiazole-4-carboxylate hydrobromide

Inchi Code

1S/C6H8N2O2S.BrH/c1-2-10-5(9)4-3-11-6(7)8-4;/h3H,2H2,1H3,(H2,7,8);1H

InChI key

KQLKNNNNKWTAMQ-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Room Temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/127942-30-7.html

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.ambeed.com/products/127942-30-7.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service