Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH98C8DC95

Ambeed, Inc.

1-(4-(5-Bromopyridin-2-yl)piperazin-1-yl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
284.16
MDL number:

Iupac Name

1-acetyl-4-(5-bromo-2-pyridinyl)piperazine

Inchi Code

1S/C11H14BrN3O/c1-9(16)14-4-6-15(7-5-14)11-3-2-10(12)8-13-11/h2-3,8H,4-7H2,1H3

InChI key

QHIUYPWBDOIIAF-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Room Temperature

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/494771-76-5.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.ambeed.com/products/494771-76-5.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service