Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH9884CC67

Ambeed, Inc.

N-Methyl-1-(6-(trifluoromethyl)pyridin-3-yl)methanamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$78.20
250 MG
$131.10
1 G
$354.20

Vendor SKU: A657648-100MG

$78.20


Availability
Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$78.20
250 MG
$131.10
1 G
$354.20

About This Item

CAS Number:
Molecular Weight:
190.17
MDL number:

Vendor SKU: A657648-100MG

$78.20


Availability
Ships in 7 days - from Aldrich Partner.

Iupac Name

N-methyl[6-(trifluoromethyl)-3-pyridinyl]methanamine

Inchi Code

1S/C8H9F3N2/c1-12-4-6-2-3-7(13-5-6)8(9,10)11/h2-3,5,12H,4H2,1H3

Inchi Key

DNSJZPZEMZAKIF-UHFFFAOYSA-N

Storage temp.

Keep in dark place,Inert atmosphere,2-8C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Liquid or Solid or Semi-solid or lump

Product Link

https://www.ambeed.com/products/886364-80-3.html

pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.ambeed.com/products/886364-80-3.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service