Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH9884C4B0

Ambeed, Inc.

3-Methyl-5-vinylpyridine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$63.25
250 MG
$104.65

Vendor SKU: A339039-100MG

$63.25


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
$63.25
250 MG
$104.65

About This Item

CAS Number:
Molecular Weight:
119.17
MDL number:

Vendor SKU: A339039-100MG

$63.25


Ships in 14 days - from Aldrich Partner.

Iupac Name

3-methyl-5-vinylpyridine

Inchi Code

1S/C8H9N/c1-3-8-4-7(2)5-9-6-8/h3-6H,1H2,2H3

InChI key

WIAMCQRXSYEGRS-UHFFFAOYSA-N

storage temp.

Sealed in dry,2-8C

Purity

97%

Country of Origin

CN

Shipping Temperature

Cold-chain transportation

Physical Form

liquid

Product Link

https://www.ambeed.com/products/51961-51-4.html

pictograms

Flame

Signal Word

Danger

Hazard Statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service