Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH9884B8E5

Ambeed, Inc.

2-[4-(Difluoromethoxy)phenyl]ethan-1-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
187.19
MDL number:

Iupac Name

2-[4-(difluoromethoxy)phenyl]ethylamine

Inchi Code

1S/C9H11F2NO/c10-9(11)13-8-3-1-7(2-4-8)5-6-12/h1-4,9H,5-6,12H2

InChI key

QVANIQIFGOVCFT-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,2-8C

Purity

95%

Country of Origin

CN

Shipping Temperature

Cold-chain transportation

Physical Form

liquid

Product Link

https://www.ambeed.com/products/771581-14-7.html

Synonym

2-(4-(Difluoromethoxy)phenyl)ethan-1-amine

pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.ambeed.com/products/771581-14-7.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service