Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH5806483B

Ambeed, Inc.

1-Chloro-4-fluoro-2-methylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
144.58
MDL number:

Iupac Name

1-chloro-4-fluoro-2-methylbenzene

Inchi Code

1S/C7H6ClF/c1-5-4-6(9)2-3-7(5)8/h2-4H,1H3

InChI key

NIOGDCDTKPQEAT-UHFFFAOYSA-N

storage temp.

Sealed in dry,Room Temperature

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.ambeed.com/products/33406-96-1.html

pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service