Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AMBH3155FD4B

Ambeed, Inc.

Ethyl Allyl(2-oxoethyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
171.2
MDL number:

Iupac Name

ethyl allyl(2-oxoethyl)carbamate

Inchi Code

1S/C8H13NO3/c1-3-5-9(6-7-10)8(11)12-4-2/h3,7H,1,4-6H2,2H3

InChI key

IQQODTFRDRREIO-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Store in freezer, under -20C

Purity

98%

Country of Origin

CN

Shipping Temperature

Shipped in cold pack

Physical Form

liquid

Product Link

https://www.ambeed.com/products/128740-03-4.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.ambeed.com/products/128740-03-4.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service