Iupac Name
methyl 2-(3-(4-hydroxyphenyl)-2,5-dihydroisoxazol-5-yl)acetate
Inchi Code
1S/C12H13NO4/c1-16-12(15)7-10-6-11(13-17-10)8-2-4-9(14)5-3-8/h2-6,10,13-14H,7H2,1H3
InChI key
SBWGRTBDBLTLPD-UHFFFAOYSA-N
storage temp.
Keep in dark place,Sealed in dry,Store in freezer, under -20C
Purity
98%
Country of Origin
CN
Shipping Temperature
Shipped in cold pack
Physical Form
solid
Product Link
https://www.ambeed.com/products/478336-92-4.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/478336-92-4.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service