AMBH2D6FB2AE
Ambeed, Inc.
(3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl)methanamine hydrochloride
Select a Size
Vendor SKU: A421582-100MG
$49.45
Select a Size
About This Item
Vendor SKU: A421582-100MG
$49.45
Iupac Name
(3,4-dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl)methanamine hydrochloride
Inchi Code
1S/C11H15NO2.ClH/c1-13-10-4-7-3-8(6-12)9(7)5-11(10)14-2;/h4-5,8H,3,6,12H2,1-2H3;1H
InChI key
UWGJJMDXKWGAED-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
Precautionary statements
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service