Key Documents
Z692239
Select autoclave tape
roll W × L 19 mm × 50 m
About This Item
UNSPSC Code:
41100000
packaging
pkg of 1 ea
roll W × L
19 mm × 50 m
General description
Autoclavable tape indicates when steam sterilisation has occurred by means of a visible black strip which shows after autoclaving (3 mins @ 134°C, 15 mins @ 121°C) Good adhesion and clear indication.
Rolls individually wrapped.
Choose from one of the most recent versions:
Certificates of Analysis (COA)