Skip to Content
MilliporeSigma

511366

Tris(tetramethylcyclopentadienyl)gadolinium(III)

99.9% trace metals basis

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H39Gd
CAS Number:
Molecular Weight:
520.85
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12352103
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

99.9% trace metals basis

form

powder or crystals

reaction suitability

core: gadolinium, reagent type: catalyst

SMILES string

[Gd].C[C]1[CH][C](C)[C](C)[C]1C.C[C]2[CH][C](C)[C](C)[C]2C.C[C]3[CH][C](C)[C](C)[C]3C

InChI

1S/3C9H13.Gd/c3*1-6-5-7(2)9(4)8(6)3;/h3*5H,1-4H3;

InChI key

HJKCHNDMWJQGKU-UHFFFAOYSA-N

General description

Atomic number of base material: 64 Gadolinium


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Sol. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 3

flash_point_f

140.0 °F - closed cup

flash_point_c

60 °C - closed cup

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library