Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC7H30368C5A

Advanced Chemical Intermediates Ltd.

5-(2-Trifluoromethoxy-phenyl)-furan-2-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
256.18
MDL number:

Iupac Name

5-[2-(trifluoromethoxy)phenyl]-2-furaldehyde

Inchi Code

1S/C12H7F3O3/c13-12(14,15)18-11-4-2-1-3-9(11)10-6-5-8(7-16)17-10/h1-7H

Inchi Key

UTGWIIGCXFJJHZ-UHFFFAOYSA-N

Storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Product Link

https://www.acints.com/products/ACI-04955

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.acints.com/products/ACI-04955


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service