Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC7508834735

Advanced Chemical Intermediates Ltd.

3-(4-Fluoro-phenoxy)-5-trifluoromethyl-pyridine-2-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
282.2
MDL number:

Iupac Name

3-(4-fluorophenoxy)-5-(trifluoromethyl)-2-pyridinecarbonitrile

Inchi Code

1S/C13H6F4N2O/c14-9-1-3-10(4-2-9)20-12-5-8(13(15,16)17)7-19-11(12)6-18/h1-5,7H

InChI key

KEYZTFPYVLCXAZ-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Product Link

https://www.acints.com/products/ACI-03982

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.acints.com/products/ACI-03982


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service