Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ADVH98DE64F3

AChemBlock

4-chloro-8-isopropyl-2-methylsulfanyl-pyrazolo[1,5-a][1,3,5]triazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
242.73
MDL number:

Iupac Name

4-chloro-8-isopropyl-2-(methylthio)pyrazolo[1,5-a][1,3,5]triazine

Inchi Code

1S/C9H11ClN4S/c1-5(2)6-4-11-14-7(6)12-9(15-3)13-8(14)10/h4-5H,1-3H3

InChI key

RRBPEHJNXHJYQN-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

0.97

Country of Origin

CN

Physical Form

light yellow solid

Product Link

https://www.achemblock.com/w165315-4-chloro-8-isopropyl-2-methylsulfanyl-pyrazolo-1-5-a-1-3-5-triazine.html

Synonym

4-chloro-8-isopropyl-2-(methylthio)pyrazolo[1,5-a][1,3,5]triazine

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/w165315-4-chloro-8-isopropyl-2-methylsulfanyl-pyrazolo-1-5-a-1-3-5-triazine.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service