Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ADVH7F380FE0

AChemBlock

2-(3-Boc-aminophenoxy)ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$184.00
1 G
$448.50
5 G
$1,794.00

Vendor SKU: P37645-250MG

$184.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$184.00
1 G
$448.50
5 G
$1,794.00

About This Item

CAS Number:
Molecular Weight:
253.3
MDL number:

Vendor SKU: P37645-250MG

$184.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

tert-butyl 3-(2-hydroxyethoxy)phenylcarbamate

Inchi Code

1S/C13H19NO4/c1-13(2,3)18-12(16)14-10-5-4-6-11(9-10)17-8-7-15/h4-6,9,15H,7-8H2,1-3H3,(H,14,16)

InChI key

AJOLDYXZBCUERL-UHFFFAOYSA-N

storage temp.

Store at Room Tem.

Purity

0.95

Country of Origin

CN

Product Link

https://www.achemblock.com/p37645-2-3-boc-aminophenoxy-ethanol.html

Synonym

tert-butyl (3-(2-hydroxyethoxy)phenyl)carbamate

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/p37645-2-3-boc-aminophenoxy-ethanol.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service