Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ADV390218025

AChemBlock

Pyrrolo[1,2-b]pyridazin-4-ylamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$207.00
100 MG
$534.75
250 MG
$891.25
1 G
$2,231.00
5 G
$6,698.75

Vendor SKU: G-6647-25MG

$207.00


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
25 MG
$207.00
100 MG
$534.75
250 MG
$891.25
1 G
$2,231.00
5 G
$6,698.75

About This Item

CAS Number:
Molecular Weight:
133.15
MDL number:

Vendor SKU: G-6647-25MG

$207.00


Ships in 10 days - from Aldrich Partner.

Iupac Name

pyrrolo[1,2-b]pyridazin-4-ylamine

Inchi Code

1S/C7H7N3/c8-6-3-4-9-10-5-1-2-7(6)10/h1-5H,8H2

InChI key

RZDNUSWOUZKGEC-UHFFFAOYSA-N

storage temp.

Store at 0-8 C

Purity

0.97

Country of Origin

CN

Physical Form

black solid

Product Link

https://www.achemblock.com/g-6647-pyrrolo-1-2-b-pyridazin-4-ylamine.html

Synonym

pyrrolo[1,2-b]pyridazin-4-amine

pictograms

Corrosion

Signal Word

Danger

Hazard Statements

MSDS

https://www.achemblock.com/g-6647-pyrrolo-1-2-b-pyridazin-4-ylamine.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service