Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AC5424074011

Accela ChemBio Inc.

4-Benzyloxy-3-nitrobenzoic Acid

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
273.25
MDL number:

Iupac Name

4-(benzyloxy)-3-nitrobenzoic acid

Inchi Code

1S/C14H11NO5/c16-14(17)11-6-7-13(12(8-11)15(18)19)20-9-10-4-2-1-3-5-10/h1-8H,9H2,(H,16,17)

InChI key

ZPGYWCMZSUFFFM-UHFFFAOYSA-N

storage temp.

+4C

Purity

95%

Country of Origin

CN

Shipping Temperature

RT

Physical Form

White to yellow solid

Product Link

http://www.accelachem.com/cn/search.html?keyword=SY013956&attname=GoodsCode

MSDS

http://www.accelachem.com/cn/msds/SY013956.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service