AC5414275714
Accela ChemBio Inc.
N'-Isopropylisonicotinohydrazide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
N'-isopropylisonicotinohydrazide
Inchi Code
1S/C9H13N3O/c1-7(2)11-12-9(13)8-3-5-10-6-4-8/h3-7,11H,1-2H3,(H,12,13)
InChI key
NYMGNSNKLVNMIA-UHFFFAOYSA-N
storage temp.
+4C
Purity
97%
Country of Origin
CN
Shipping Temperature
RT
Physical Form
White to yellow solid
Product Link
http://www.accelachem.com/cn/search.html?keyword=SY025980&attname=GoodsCode
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
http://pdf.accelachem.com/cn/msds/SY025980.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service